Learn More
Rogo Sampaic™ Borosilicate Glass Coverslips
Click to view available options
:
Size: 24L x 32mmW
Specifications
Specifications
| Product Type | Coverslip |
| Shape | Rectangle |
| Material | Borosilicate Glass |
| Length (Metric) | 24 mm |
| Width (Metric) | 32 mm |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Quantity | 100/Pk. |