Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
Click to view available options
Standard Taper Joint No.:
Socket: 19/26
Socket: 29/32
Socket: 45/40
Cone Size:
14/23
19/26
24/29
29/32
45/50
Unit Size:
Each
Description
- Solid rim
- Acc. to DIN 12257
- Reinforced ground joint
Specifications
Specifications
| Cone Size | 14/23 |
| Material | Borosilicate Glass |
| Product Type | Duran sleeve |
| Quantity | 1 Each |
| Standard Taper Joint No. | Socket: 19/26 |