Learn More
Duran™ Borosilicate Glass Erlenmeyer Flask with 29/32 NS Standard Ground Joint
150.00€ - 228.00€
Specifications
Material Borosilicate Glass
Shape Conical
View More Specs
Products 6
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Certifications/Compliance Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Certifications/Compliance Price Quantity & Availability  
15251889 View Documents Duran™
241932705
100 mL 64 mm 105 mm ISO 4794 and USP Standard 191.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
15261889 View Documents Duran™
241933204
200 mL 79 mm 131 mm USP Standard 185.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
15281889 View Documents Duran™
241933701
250 mL 85 mm 140 mm ISO 4794 and USP Standard 200.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
11850620 View Documents Duran™
241933907
300 mL 87 mm 156 mm USP Standard 190.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
11756268 View Documents Duran™
241931906
50 mL 51 mm 85 mm ISO 4794 and USP Standard 150.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
15201899 View Documents Duran™
241934603
500 mL 105 mm 175 mm ISO 4794 and USP Standard 228.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
09-06-2020
Add to basket
Description
Description
  • Easy-to-read scale and large labelling field for easy marking
  • Conical shape makes these flasks ideal for mixing liquids
  • Suitable for use as heating glassware
  • Manufactured from BoroSilicate glass
Specifications
Specifications
Borosilicate Glass
Conical