Learn More
IKA™ Borosilicate Glass Evaporating Flask For RV 10.81 Rotary Evaporators
Brand: IKA™ 0003740200
38.64 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.10000kg
Specifications
Borosilicate Glass | |
RV 10.81 Rotary Evaporators | |
100 mL |
Evaporating Flask | |
1 Each |