Learn More
BRAND™ BISTABIL Lateral Stopcock
Borosilicate glass construction. Includes interchangeable borosilicate glass key with retention device.
Brand:  BRAND™ 81809
Additional Details : Weight : 0.03000kg
Product Code. 10309621
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Borosilicate glass (Boro 3.3) construction
  • Stopcock housing with precision Boro 3.3 burette tip
  • Interchangeable borosilicate glass key with retention device
  • Stopcock size 3NS/12
Specifications
Specifications
50 mL
Borosilicate Glass
Retention Device
Burette Cap
1 Each
Stopcock