Learn More
Witeg™ Borosilicate Glass Laboratory Bottles, Wide Mouth
Supplement your experimentation using the Witeg™ Borosilicate Glass Laboratory Bottles, Wide Mouth for your laboratory needs.
Brand:  Witeg™ 5526002S
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11803871
  • 112.00€ / Pack of 10
Estimated Shipment
14-06-2021
Add to basket
Description
Description
  • Wide Moutn
  • Glass
  • 45 Thread
Specifications
Specifications
GL 45
2000 mL
Borosilicate Glass
10Pack
Yes
Screw cap and pouring ring
10
Laboratory Bottle