Learn More
Soda Lime Glass Petri Dishes
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
60 mm
80 mm
100 mm
120 mm
150 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Each
Pack of 18
Pack of 20
Description
- Constructed of soda lime glass.
- Includes lid
Specifications
Specifications
| Closure Included | Closure Included |
| Material | Soda Lime Glass |
| Shape | Round |
| Type | Petri Dish |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 120 mm |