Learn More
Witeg™ Borosilicate Glass Powder Funnel, 100 to 110 mm
Witeg™ Borosilicate Glass Powder Funnels are available in a variety of cone tapers, angles and outer diameters.
Brand:  Witeg™ 1 129 030
Additional Details : Weight : 0.80000kg
Product Code. 11857800
  • 8.70€ / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass construction
  • Available in a variety of tapers, angles, and diameters
Specifications
Specifications
Funnel
100 to 110 mm
Borosilicate Glass
40°