Learn More
Witeg™ Borosilicate Glass Powder Funnel with ST Cone
Product Code. p-8010326
Click to view available options
Diameter (Metric) Stem:
80 mm
100 mm
150 mm
Cone Size:
19/26
24/29
29/32
Unit Size:
Each
Pack of 10
Product Code. 11867680
Brand: Witeg™ 1 650 019
Only null left
Add to basket
Add to basket
Witeg™ Borosilicate Glass Powder Funnels are available in a variety of cone tapers, angles and outer diameters.
  • ST cone
  • Borosilicate glass construction
  • Available in a variety of tapers, angles, and diameters
Specifications
Diameter (Metric) Stem 80 mm
Material Borosilicate Glass
Angle 60°
Cone Size 19/26
Product Type Funnel