Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Product Code. 10694192
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
19/26 NS
29/32 NS
Pore Size:
2.5mm
4mm
6mm
Unit Size:
Pack of 2
Product Code. 10694192
Brand: Lenz Laborglasinstrumente™ 04030158
Only null left
Add to basket
Add to basket
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • With inscription label
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Joint Number 29/32 NS
Porosity Yes
Pore Size 4mm
Stopper Material Polyethylene