Learn More
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap
Click to view available options
Volume (Metric):
1 mL
4 mL
24 mL
Unit Size:
Pack of 144
Description
- Plain top design
- Lab pack quantities
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Specifications
| Volume (Metric) | 1 mL |
| No. per Case | 1440 |
| No. per Pack | 144 |