Learn More
Kimble Chase™ Borosilicate Glass Shells Vials
Click to view available options
Volume (Metric):
2 mL
44 mL
Unit Size:
Pack of 144
Specifications
Specifications
| Volume (Metric) | 2 mL |
| No. per Case | 2304 |
| No. per Pack | 144 |
| Closure Type | Open Bottom Plug |