Learn More
Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Pack of 4
Description
- Heavy wall construction for increased mechanical strength
- Much more robust than standard beakers
- Full 3.3 expansion borosilicate glass
- Reinforced rim with 'easy pour' pouring spout
- Graduated in white ceramic with marking area
- Uniform wall thickness for optimum optical clarity
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Includes | Spout |
| Quantity | 10 |
| Type | Beaker |
| Graduated | Yes |
| Scale | White Ceramic Scale |
| Style | Squat Form |