Learn More
Fisherbrand™ Borosilicate Glass Tall Form Beakers
Product Code. 11902233 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
800 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 10
Product Code. 11902233
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beakers are available in a variety of sizes.
  • Borosilicate glass
  • Tall form
  • Graduated
  • With spout
  • Manufactured to ISO 3819, DIN 12331
TRUSTED_SUSTAINABILITY
Specifications
Color Translucent
Diameter (Metric) Outer 120 mm
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Finish Matte
Height (Metric) 240 mm
Includes Spout
Quantity 10 Pk.
Type Beaker
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Scale White Graduated Scale
Style Tall Form