Learn More
IKA™ Borosilicate Glass Uncoated Receiving Flask
Shop All IKA Products
Click to view available options
Volume (Metric):
250 mL
500 mL
2000 mL
3000 mL
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass |
| Type | Receiving Flask |
| For Use With (Equipment) | RV 10.105 Evaporator |
| Quantity | 1 Each |
| Volume (Metric) | 3000 mL |