Learn More
B2M™ Bottle Brushes
Product Code. 12078758
Click to view available options
Length (Metric) Handle:
190 mm
235 mm
250 mm
260 mm
270 mm
300 mm
400 mm
440 mm
450 mm
500 mm
600 mm
Length (Metric) Brush:
50 mm
60 mm
75 mm
80 mm
90 mm
100 mm
110 mm
120 mm
Diameter (Metric) Bristle:
4 mm
5 mm
7 mm
8 mm
10 mm
12 mm
13 mm
15 mm
18 mm
20 mm
25 mm
30 mm
50 mm
55 mm
Tip Type:
2 Wicks
Bound Bristle Tip
Cotton Tip
Return End
Shell End
Without End
Without Tip
Unit Size:
Pack of 10
Product Code. 12078758
Brand: B2M™ 10095
Only null left
Add to basket
Edge
Add to basket
Clean all types of bottles and tubes. B2M™ Bottle Brushes are available with silk, nylon or goat hair bristles for the full range of uses. Brush styles and sizes are available for efficient cleaning of any size or type of receptacle.
  • Three choices of bristles:
    • White silk bristles for general cleaning use
    • Nylon bristles are very resistant for use with aggressive substances
    • Goat hair bristles are very soft for delicate cleaning
  • A variety of tip styles, including: bound-bristle, 2 wicks, cotton and shell tips
  • For cleaning all receptacles: tubes, burettes, balls and bottles
Specifications
Product Type Bottle Brush
Bristle Material Silk
Tip Type Return End
Diameter (Metric) Bristle 50 mm
For Use With (Application) General purpose cleaning
For Use With (Equipment) Ball
Length (Metric) Brush 90 mm
Length (Metric) Handle 440 mm
Quantity 10 Pk.