Learn More
Wuertt Celluloid™ Bottle Carrier
Click to view available options
Holds:
Bottles (1 L)
Bottles (250 or 500 mL)
Positions:
6
10
Unit Size:
Each
Specifications
Specifications
| Product Type | Bottle Rack |
| Holds | Bottles (250 or 500 mL) |
| Positions | 10 |
| Shelves | 10 |
| Quantity | 1 Each |