Learn More
Corning™ Bottle Top Dispensers (BTD) Outer Housing
Product Code. 16882090 Shop All Corning Products
Click to view available options
Volume (Metric):
2.5 mL
5 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Product Code. 16882090
Brand: Corning™ 6849
Only null left
Add to basket
Add to basket
Outer housing for BTD and dispensing reagents
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 50 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters