Learn More
Amsta Rostfritt Stainless Steel Bottle Without Lid With Skirtband
Click to view available options
Capacity (Metric):
4 L
5 L
6 L
8 L
10 L
Unit Size:
Each
Description
- Stainless steel 304 /1.4301
Specifications
Specifications
| Product Type | Pail |
| Capacity (Metric) | 4 L |
| Color | Silver |
| Material | Stainless Steel |