Learn More
Fisher BioReagents™ Bovine Serum Albumin Powder, Fraction V
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| pH Range | 7pH |
| Quantity | 100 g |
| Product Type | Bovine Serum Albumin Powder |
Safety and Handling
Product Identifier
- Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0
- Warning
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.