Learn More
Trajan Scientific and Medical™ BP1 GC Capillary Columns
Product Code. 15258633
Click to view available options
Length (Metric):
15 m
25 m
30 m
50 m
60 m
Diameter (Metric):
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
5 μm
Unit Size:
Each
Product Code. 15258633
Brand: Trajan Scientific and Medical™ 054068
Only null left
Add to basket
Add to basket
Suitable for all routine analysis. Trajan Scientific and Medical BP1 GC Capillary Columns are industry standard non polar phase columns.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Diameter (Metric) 0.32 mm
Type BP1 GC Column
For Use With (Application) Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
Temperature Range -60°C to +340/360°C
Film Thickness 0.5 μm
Length (Metric) 30 m
Material 100% Dimethyl Polysiloxane
Packing Material BP1