Learn More
Trajan™ BPX Volatiles Column
Product Code. p-4824114
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
1.4 μm
1.8 μm
3 μm
Unit Size:
Each
Product Code. 12706567
Brand: Trajan™ 054984
Only null left
Add to basket
Add to basket
SGE BPX Volatiles Column is a polar phase designed for EPA volatile organics analysis
  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Max. Temperature 300°C
Diameter (Metric) Inner 0.53 mm
Stationary Phase Cyanopropylphenyl Polysiloxane
USP Type G43
For Use With (Application) Environmental Analysis, Volatile Organic Compounds Analysis, Alcohol Analysis
Temperature Range 0°C to 290/300°C
Film Thickness 3 μm
Length (Metric) 30 m
Packing Material BPX-Volatiles
Polarity Polar
Coating Fused Silica