Learn More
Brady™ BBP™ Hi-Performance Polyester Tape
BBP™ Hi-Performance Polyester Tape (B-569) has low halide content and is designed for marking stainless steel pipes particularly in harsh environments.
Brand:  Brady™ 117975
 View more versions of this product
Product Code. 11726435
  • ONLINE EXCLUSIVE PRICE
    244.00€
    223.28€ / Pack of 1
    Save 20.72€
    8% Off
In Stock
Description
  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester tape can be thermal transfer printed
  • Withstands mild chemicals, oil and water
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Specifications
Specifications
High Performance Polyester Tape for BBP3x Printers
Red
Tape
Thermal Transfer
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 212°F
45 lb./in.
0.076 mm
30.48 m
29 mm
Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Label Tape
Designed for use on Stainless Steel pipes in harsh environments such as the pulp and Paper industry
80 Gardner Units
3 to 5 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +100°C
0.003 in.
100 ft.
1.12 in.
B30C-1125-569-RD