Learn More
Brady™ BBP™ Tamper-Evident Silver Metallized Polyester Label
Product Code. 11731795
Click to view available options
Height (English):
2 in.
3 in.
Length (English):
1 in.
2 in.
100 ft.
Unit Size:
Each
Product Code. 11731795
Brand: Brady™ 118076
Only null left
Add to basket
Add to basket
BBP™ Tamper-Evident Silver Metallized Polyester Labels B-438 have a permanent adhesive that leaves a checkerboard footprint if removed.
  • Designed for rating and serial plates requiring both high-performance and protection against removal
  • Ideal for product labeling, asset tracking and general industrial ID
  • Withstands solvents and variable temperatures
  • Requires minimal handling in order to prevent prematurely exposed checkerboard pattern
Specifications
Certifications/Compliance RoHS 2002/95/EC, UL969
Description Tamper-Evident Metallised Labels for BBP3x Printers
Adhesive Type Acrylic, Tamper-evident
Coating None
Color Silver
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For asset tracking, general ID
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1225 Labels/Box
Quantity 1 Roll
Shape Rectangle
Software Type Brady Workstation, LabelMark, MarkWare
Temperature Range (English) -40°F to +100°F
Temperature Range (Metric) -40°C to +37°C
Tensile Strength 60 lb./in.
Thickness (English) 0.0033 in.
Thickness (Metric) 0.084 mm
Vertical Repeat (Metric) 28.60 mm
Dimensions (LxW) 2 x 1 in.
Length (English) 1 in.
Length (Metric) 2.54 cm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 2 in.
Height (Metric) 5.08 cm
Alternate Part Number B30-17-438