Learn More
Brady™ Label Maker Vinyl Cartridges (B-595) for BMP51, BMP53, BMP41 Printers
Product Code. 15153359
Click to view available options
Color:
Black, Orange
Black, White
Black, Yellow
Red, White
White, Black
White, Blue
White, Green
White, Red
Unit Size:
Each
Product Code. 15153359
Brand: Brady™ 131598
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Indoor/Outdoor grade vinyl conforms well to smooth or rough surfaces
  • Durable and low-shrink with aggressive adhesive
  • Used on surfaces such as pipes, walls, doors, panels, shelving, bins and equipment
  • Offers resistance to oils, detergents, dirt, grime and many solvents
Specifications
Color White, Green
Material Vinyl (B-595)
Product Type Label Maker Cartridge
For Use With (Equipment) BMP™51,BMP™53,BMP™41 Printers
Quantity 1 Ea.
Size (English) 25 ft. x 1 in.
Size (Metric) 7.62 m x 25.4 mm
Length (English) 25 ft.
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (Metric) 7.62 m