Learn More
Brady™ LABXPERT™ Printer Labels
Product Code. p-3649395
Click to view available options
Quantity:
150/Pk.
175/Pk.
Height (Metric):
1.52 cm
5.58 cm
5.71 cm
Material:
Freezerbondz I Polyester
Self-laminating Polyester
Width (English):
1.5 in.
2.62 in.
Unit Size:
1 roll
Each
Product Code. 10153042
Brand: Brady™ 060905
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specialty laboratory labels designed for use with these specific label printers
  • Printable, self-laminating high-performance laboratory identification for LABXPERT printer.
  • Polyester label withstands temps -196° to +121°C
Ultrathin construction prevents jamming in racks & centrifuge
Specifications
Description Surface Mount
Adhesive Type Acrylic, Permanent
Color Black, White
Finish Matte
Format Cartridge (Label)
Material Freezerbondz I Polyester
Product Type Laboratory Label
For Use With (Application) Conical tube, laboratory identification, tube
For Use With (Equipment) LabXpert™ Printer
Quantity 175/Pk.
Shape Rectangle
Temperature Range (English) -320.8 to 249.8°F
Temperature Range (Metric) -196°C to +121°C
Thickness (Metric) 0.053 mm
Width (English) 1.5 in.
Width (Metric) 38.10 mm
Height (English) 2.2 in.
Height (Metric) 5.58 cm