Learn More
Brady™ ToughStripe™ Floor Marking Prespaced Marks
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Shape:
Rectangle
Round
Unit Size:
1 roll
Description
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Tape removes cleanly without any unsightly adhesive residue or damage to the floor
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.36
Specifications
Specifications
| Certifications/Compliance | ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22 |
| Description | ToughStripe Prespaced shapes |
| Adhesive Type | Rubber, Permanent |
| Coating | None |
| Color | Blue |
| Finish | Gloss |
| Format | Tape |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | None |
| Show More |