Learn More
Brady™ BradyJet™ Continuous Vinyl Labels
Brady™ BradyJet™ Continuous Vinyl Labels are ideal for a wide range of identification applications.
Marca: Brady™ 150010
Descrizione
- Constructed of indoor-use general purpose vinyl
- Features a specially-formulated topcoat for improved durability and color
- Specially-formulated topcoat enhances colors and label appearance
- Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
- Topcoat protects print from smudging and harsh conditions
- Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifica
| Acrylic, Permanent | |
| White | |
| Roll | |
| Single-side Printable | |
| Label | |
| Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels | |
| 1 | |
| 1 Tape/Bag | |
| -40°C to +60°C | |
| 0.135 mm | |
| 100 ft. | |
| 1.125 in. | |
| 1.88 in. I.D. |
| None | |
| Matte | |
| Vinyl | |
| Ink Jet | |
| Non-sterile | |
| BradyJet™ J2000 Printer | |
| -40°C | |
| -40°F to +140°F | |
| 0.0053 in. | |
| 100 ft. x 1.125 in. | |
| 30.48 m | |
| 28.58 mm | |
| J20C-1125-2595 |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.