Learn More
Brady™ BradyJet™ J5000 Repositionable Inkjet Vinyl Labels
Product Code. 17429212
Click to view available options
Dimensions (LxW):
100 ft. x 2.25 in.
100 ft. x 4 in.
100 ft. x 6 in.
100 ft. x 8 in.
Unit Size:
Each
Product Code. 17429212
Brand: Brady™ 151112
The special aggressive adhesive on this Repositionable Inkjet Material stays in place when you need it to, but removes cleanly without leaving adhesive residue making it ideal for 5S and inventory applications.
  • Durable and flexible material making it ideal for areas where identification is frequently changed
  • Service temperature of -40 to 140°F (-40 to 60°C)
Specifications
Adhesive Type Acrylic, Removable
Coating None
Color White
Display Blank
Finish Matte
Format Continuous
Material Vinyl
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Temporary Labeling
For Use With (Equipment) BradyJet™ J5000
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Label/Bag
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.01 in.
Thickness (Metric) 0.254 mm
Dimensions (LxW) 100 ft. x 6 in.
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 6 in.
Width (Metric) 152.40 mm
Alternate Part Number J50C-6000-2581