Learn More
HUBER Braided Hose
Product Code. p-8013458
Click to view available options
For Use With (Equipment):
Cooling Water PZ-100-2-G1 1/4 in.
Cooling Water PZ-90-1-G1 1/4
Cooling Water PZ-90-1.5-G1 1/4
Unit Size:
Each
Product Code. 11922712
Brand: HUBER 18022
Only null left
Add to basket
Add to basket
For cooling water
  • Braided hose
  • Min. temp.: -20°C
  • Max. temp.: 90°C
  • Connection: 1 1/4 inch
Specifications
For Use With (Equipment) Cooling Water PZ-90-1.5-G1 1/4
Temperature Range -20°C to +90°C
Type Braided Hose