missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 384-Well, cellGrade-Treated, Flat-Bottom Microplate

Catalog Number p-9045023
Click to view available options
Packaging:
5/Pk., 32/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Certifications/Compliance ISO 11137, DIN EN ISO 10993
Lid With Lid
Packaging 5/Pk., 32/Cs.
Color White, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment cellGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of adherent cell lines
No. per Pack 5
Quantity 32/Cs.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'