missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pointes à filtre BRAND™ BIO-CERT™, 50 à 1000 µl

En PP haute pureté sans diHEMDA ni oléamide. Les pointes à filtre BrandTechTM BRANDTM BIO-CERTTM de 50 à 1000 µl sont stériles et ne contiennent aucun contaminant biologique. Les filtres en PE ne sont pas auto-étanches et permettent la récupération des échantillons par filtres humides. Paroi fine, translucide, éjection facile et réduction des pertes. Graduations à 250, 500 et 1000 µl. Disponibles en rétention standard ou ultra-faible.

Marque:  BRAND™ 732834

Informations supplémentaires : Poids : 1.71000kg

 Afficher plus de versions de ce produit

Code produit. 15104997

  • 120.00€ / 960 pièces


Expédition estimée
17-06-2020
Ajouter au panier

Description

Description

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Non self-sealing PE filters allow sample recovery with wetted filters
  • Thin-walled for high translucency, easy ejection, and low waste.
  • Graduations at 250, 500, and 1000μL for quick volume check.
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format; racked tips are colorless and placed into blue-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties

Spécification

Spécification

Rack
50 à 1000 μl
Stérile
250, 500, 1000 μl
Polypropylène
DNase, RNase, ATP, and Endotoxin Free
Rétention ultra faible
Filtered
Incolore
Oui
7 cm
Vidéos
Documents