Learn More
BRAND™ BRANDplates immunoGrade™ 96-Well Microplates
Product Code. 10477863 Shop All Brand Products
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
Product Code. 10477863
Brand: BRAND™ 781724
Only null left
Add to basket
Add to basket
An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays
  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color White
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment immunoGrade
Total Volume (Metric) Well 330 μL
Well Shape Round
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Quantity 100/Cs.