Learn More
BRAND™ Filter Membranes
Used with seripettor™ / QuikSip™ / accu-jet™ pro macro pipet controllers. Filter Membranes are available in 3μL capacity and provided in blister pack packaging.
Brand: BRAND™ 26052
7.56 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.04500kg
Description
- For seripettor™ / QuikSip™ / accu-jet™ pro / macro pipette controller
- Blister pack packaging
- 3μm
- Non-sterile
Specifications
Polypropylene, PTFE | |
3 μm | |
Reduces cross-contamination, protects against over aspiration and allows pipetting to Class A tolerances | |
Macro and Accu-jet™ pipet controllers |