Learn More
VACUUBRAND™ Quick Coupling for BVC Fluid Aspiration Systems
Replacement parts and accessories for VACUUBRAND BVC fluid aspiration systems
Marque:  VACUUBRAND™ 20635808
Informations supplémentaires : Weight : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 12615650
  • 113.00€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Ajouter au panier
Description
Description
The BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work. They provide the flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in the bottle.
635807
  • Quick coupling from VHC to collection vessel
635808
  • Quick coupling from collection vessel to pump
Spécification
Spécification
BVC fluid aspiration system
Quick coupling from collection vessel to pump