Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Support Racks
Product Code. 10204634
Click to view available options
For Use With (Equipment):
Ultrasonic Bath, Model 1800
Ultrasonic Bath, Model 2800
Ultrasonic Bath, Model 3800
Ultrasonic Bath, Model 5800
Ultrasonic Bath, Model 8800
Height (English):
8 in.
12 in.
15 in.
20 in.
Width (English):
6 in.
8 in.
15 in.
Unit Size:
Each
Product Code. 10204634
Brand: Branson Ultrasonics™ CPN916042
Only null left
Add to basket
Add to basket
For use with Branson ultrasonic baths
  • Choice of sizes to fit all baths
Specifications
Type Support Rack
For Use With (Equipment) Ultrasonic Bath, Model 5800
For Use With (Application) For Model 5800
Height (English) 15 in.
Height (Metric) 38.1 cm
Width (English) 15 in.