Learn More
Branson Ultrasonics™ Sonifier™ Cell Disruptor Acoustic Enclosure Accessory, Adapter
Click to view available options
:
Adapter for Acoustic Enclosure
Description
- Adapter needed for 150w sonifier cell disruptor
Specifications
Specifications
| Type | Adapter |
| For Use With (Equipment) | Sonifier Cell Disruptor, Models SLPt and SLPe |
| Wattage | 150 w |