Learn More
Branson Ultrasonics™ Microtips for Sonifiers SLPe, SLPt, 150E and 150T
Click to view available options
Volume Range (Metric):
10 to 100 mL
3 mL or less
3 to 10 mL
5 to 25 mL
Diameter (Metric):
0.22 cm
0.32 cm
0.48 cm
0.63 cm
Unit Size:
Each
Description
- Can handle volumes up to 100mL
- Working length: 4 in.
- Sound proof enclosure
Order Info
0.125 in. microtip is provided with the 150 watt units
Specifications
Specifications
| Product Type | SLP Microtip |
| Volume Range (Metric) | 5 to 25 mL |
| Diameter (English) | 0.19 in. |
| Diameter (Metric) | 0.48 cm |
| Length (English) | 4 in. |
| Length (Metric) | 10.16 cm |
| For Use With (Application) | Ideal for processing samples to the smallest diameter vessels |
| For Use With (Equipment) | SLPe, SLPt, 150E and 150T Sonifier™ |