Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers
Product Code. 13483639
Click to view available options
For Use With (Equipment):
Beaker in Ultrasonic Benchtop Cleaning Bath, 1 x 600 mL, Fits Model 1800
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 250 mL, Fits Model 2800
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 2800
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 3800
Beaker in Ultrasonic Benchtop Cleaning Bath, 3 x 250 mL, Fits Model 3800
Beaker in Ultrasonic Benchtop Cleaning Bath, 4 x 600 mL, Fits Model 5800
Beaker in Ultrasonic Benchtop Cleaning Bath, 6 x 600 mL, Fits Model 8800
Unit Size:
Each
Product Code. 13483639
Brand: Branson Ultrasonics™ CPN246013
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with Branson ultrasonic cleaning baths
  • Helps positon beakers securely in bath
Specifications
Type Beaker Positioning Cover
For Use With (Equipment) Beaker in Ultrasonic Benchtop Cleaning Bath, 4 x 600 mL, Fits Model 5800
For Use With (Application) Helps position beakers securely in bath