Learn More
Brass plate alloy 260, 6.35mm (0.25in) thick
Product Code. p-7059716
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Unit Size:
Each
Product Code. 15482187
Brand: Thermo Scientific Alfa Aesar 045182.GT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Brass plate alloy 260
Name Note 6.35mm (0.25 in.) thick
Quantity 150 x 150 mm
Odor Odorless
Form Plate