Learn More
Aalborg™ Precision Valve for Brass Flowmeter
Click to view available options
Airflow Rate:
1000 mL/min.
200 mL/min.
21500 mL/min.
2500 mL/min.
400 mL/min.
6200 mL/min.
Waterflow Rate:
12mL/min.
200mL/min.
26mL/min.
650mL/min.
6mL/min.
80mL/min.
Unit Size:
Each
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
| For Use With (Equipment) | 150 mm Gas/Liquid Flowmeter |
| Type | Standard Valve |
| Airflow Rate | 21500 mL/min. |
| Waterflow Rate | 650mL/min. |