Learn More
Greiner Bio-One BREATHseal™ Sealer
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Description
- Unique gas-permeable sealer with a wide distribution of pores for excellent gas exchange
- Available sterile and non-sterile
Specifications
Specifications
| Color | Translucent |
| Material | Rayon-Fibers Foil, Poly-coated Kraft Paper with Silicone |
| Sterility | Non-sterile |
| No. per Case | 500 |
| Product Type | BREATHseal Sealer |
| Adhesive | Acrylic |
| Thickness | 0.14 mm |
| Pierceable | Yes |
| Plate Compatibility | Cell Culture Microplate, HTS Microplate |