Learn More
WTW™ Brewery Application Package for the PhotoLab™ 6000/7000 Series Photometers
Click to view available options
For Use With (Equipment):
PhotoLab™ 6000/7000 Series Photometers
Unit Size:
Each
Description
- Uploadable test method package for use with the Photolab™ 6000 Series photometers
- The methods are uploaded to the meter via USB and are immediately activated
- The package offers standard methods according to MEBAK for measurement of a wide range of common parameters in the brewing industry (EBC)
- After first upload and activation, future updates can be simply downloaded from the WTW website
Specifications
Specifications
| Description | Uploadable Test methodology |
| Type | Alpha-acids; Anthocyanins; Beer; Wort: Color; Copper; Flavonoids, FAN, Iron, Iso-alpha-acids; Nickel, Iodine; Reduction Capacity; Phenols, TBA, Carbohydrate; Polyphenols, Vicinal Diketones |
| For Use With (Equipment) | PhotoLab™ 6000/7000 Series Photometers |
| Test Method | MEBAK |