Learn More
WTW™ Brewery Application Package for the PhotoLab™ 6000/7000 Series Photometers
GREENER_CHOICE
Product Code. 16481881
Click to view available options
For Use With (Equipment):
PhotoLab™ 6000/7000 Series Photometers
Unit Size:
Each
Product Code. 16481881
Brand: WTW™ 250214
The WTW™ Brewery Application Package for the PhotoLab™ 6000/7000 Series Photometers contains MEBAK standard photometric methods for the measurement of typical parameters in the brewing industry (EBC).
  • Uploadable test method package for use with the Photolab™ 6000 Series photometers
  • The methods are uploaded to the meter via USB and are immediately activated
  • The package offers standard methods according to MEBAK for measurement of a wide range of common parameters in the brewing industry (EBC)
  • After first upload and activation, future updates can be simply downloaded from the WTW website
TRUSTED_SUSTAINABILITY
Specifications
Description Uploadable Test methodology
Type Alpha-acids; Anthocyanins; Beer; Wort: Color; Copper; Flavonoids, FAN, Iron, Iso-alpha-acids; Nickel, Iodine; Reduction Capacity; Phenols, TBA, Carbohydrate; Polyphenols, Vicinal Diketones
For Use With (Equipment) PhotoLab™ 6000/7000 Series Photometers
Test Method MEBAK