missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Bridge Electrolyte for Ion Selective and Gas Sensitive Electrodes

Click to view available options
pH Range:
5.5 to 8
Unit Size:
Each
Description
With ion selective measuring procedures it is important to prevent the ion to be measured from entering the sample via the bridge electrolyte of the reference electrode. Especially the ions Cl-, K+ or NO3- often used in electro-chemical applications must never enter the sample via the bridge electrolyte during the measurement of these ions. Therefore, there is a special respective bridge electrolyte for every measuring method.
Specifications
Specifications
| pH Range | 5.5 to 8 |
| Type | Bridge Electrolyte |
| For Use With (Equipment) | Use with Ion selective and gas sensitive electrodes such as Ammonia(NH4+), Bromium(Br-), Cadmium(Cd2+) and many more |
| Volume (Metric) Sample | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction