missing translation for 'onlineSavingsMsg'
Learn More

Sony™ High-Gloss Black and White Thermal Paper for Ultrasound

Product Code. 11912657
Encompass
Click to view available options
Color:
Black and White
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11912657

Brand: Sony™ UPP110HG

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Type V, high gloss thermal imaging paper designed to provide optimal image quality when used with Sony™ digital and video printers. Sony™ High-Gloss Black and White Thermal Paper for Ultrasound can produce approximately 240 prints per roll.

  • Compatible with Sony Thermal Printer models UP-D898MD, UP-895MD, UP-897MD, UP-D895 and UP-D897 printers.
  • Optimal medical imaging quality
  • Replacement for item number E20022DJ
  • Approximately 240 prints per roll
  • 10 rolls per pack

Specifications

Color Black and White
Dimensions (L x W) 18 m x 110 mm
Finish High Gloss
For Use With (Equipment) UP-D898MD, UP-895MD, UP-897MD, UP-D895 and UP-D897 printers
Product Type High-Gloss Thermal Imaging Paper
Quantity 10 Rolls
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.