missing translation for 'onlineSavingsMsg'
Learn More

Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051928
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
4165-57-5
C6H5Br
162.04
MFCD00000056
QARVLSVVCXYDNA-RALIUCGRSA-N
bromobenzene-d5, bromo 2h5 benzene, 1-bromo 2 h? benzene, bromobenzene d5, benzene-d5, bromo, 1-bromo 2h5 benzene, c6d5br, bromobenzene-d5, 99.5 atom % d, bromobenzene-d5, reagent grade, ≥99 atom % d cp
2723968
[2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
4165-57-5
C6H5Br
162.04
MFCD00000056
QARVLSVVCXYDNA-RALIUCGRSA-N
bromobenzene-d5, bromo 2h5 benzene, 1-bromo 2 h? benzene, bromobenzene d5, benzene-d5, bromo, 1-bromo 2h5 benzene, c6d5br, bromobenzene-d5, 99.5 atom % d, bromobenzene-d5, reagent grade, ≥99 atom % d cp
2723968
[2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]

The labeled version of bromobenzene , used as a general reagent in palladium-catalyzed reaction and in the synthesize of Grignard reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.55 g/mL
Flash Point 51°C (124°F)
Odor Aromatic
Refractive Index 1.557
Quantity 2 g
Boiling Point 53°C (23mmHg)
UN Number UN2514
Solubility Information Insoluble in water.
IUPAC Name 1-bromo(²H₅)benzene
Formula Weight 162.06
Percent Purity 99%
Chemical Name or Material Bromobenzene-d5, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'