Learn More
Thermo Scientific Chemicals Bromocresol Green, sodium salt
Catalog Number p-3756511
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67763-24-0
C21H13Br4NaO5S
720.00
MFCD00148898
HEFSGAHJDGZCHA-UHFFFAOYNA-M
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
23675901
sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67763-24-0
C21H13Br4NaO5S
720.00
MFCD00148898
HEFSGAHJDGZCHA-UHFFFAOYNA-M
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
23675901
sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Clarity Passes Test
Infrared Spectrum Authentic
Formula Weight 720
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Color Black to Brown/Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromocresol Green, sodium salt, Pure