Learn More
Bromomethyl methyl ether, 90%, Thermo Scientific Chemicals
Catalog Number p-4813367
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
13057-17-5
C2H5BrO
124.97
JAMFGQBENKSWOF-UHFFFAOYSA-N
bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
83093
bromo(methoxy)methane
COCBr
13057-17-5
C2H5BrO
124.97
JAMFGQBENKSWOF-UHFFFAOYSA-N
bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
83093
bromo(methoxy)methane
COCBr
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.5300g/mL
Boiling Point 87.0°C
Flash Point 26°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2OCH3
Refractive Index 1.455
Quantity 5 g
Beilstein 01,582
Fieser 10,56
Specific Gravity 1.53
Formula Weight 124.97
Percent Purity 90%
Chemical Name or Material Bromomethyl methyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromomethyl methyl ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.