missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue Sodium Salt Indicator, Honeywell Fluka™

Catalog Number 15653850
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
34725-61-6
C19H9Br4NaO5S
MFCD00013793
This item is not returnable. View return policy
34725-61-6
C19H9Br4NaO5S
MFCD00013793

Indicator

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
UN Number NONH for all modes of transport
Beilstein 6995701
Solubility Information Soluble in water
Formula Weight 691.94g/mol
Packaging Glass Bottle
Color Blue
pH 4.6
Quantity 10 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue Sodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'