missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Honeywell Fluka™
39.61 EUR valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
UN Number | NONH for all modes of transport |
Beilstein | 373934 |
Formula Weight | 624.38g/mol |
Grade | ACS Reagent |
Packaging | Glass Bottle |
Suitability | Complies for clarity of solution |
Visual Transition Interval | 5.8 to 7.4 |
Color | Violet |
Quantity | 5 g |
Physical Form | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'